Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGCAATGGCAGGCGCTGGTGAG[C/T]GCAAAGGCAAGAAGGATGACAATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604064 MIM: 615497 | ||||||||||||||||||||
Literature Links: |
ATF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATF4 - activating transcription factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIEF1 - mitochondrial elongation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304564.1 | 732 | Missense Mutation | CGC,TGC | R,C 7 | NP_001291493.1 | |
NM_019008.5 | 732 | Missense Mutation | CGC,TGC | R,C 7 | NP_061881.2 | |
XM_011530236.1 | 732 | Missense Mutation | CGC,TGC | R,C 7 | XP_011528538.1 | |
XM_011530237.1 | 732 | Missense Mutation | CGC,TGC | R,C 7 | XP_011528539.1 | |
XM_011530238.1 | 732 | Missense Mutation | CGC,TGC | R,C 7 | XP_011528540.1 | |
XM_017028838.1 | 732 | Missense Mutation | CGC,TGC | R,C 7 | XP_016884327.1 |