Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGTGCAGGTACTCTGCGTGCGC[A/C]CGCTGGATCTCCTCCAGGTCGCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607917 MIM: 610053 | ||||||||||||||||||||
Literature Links: |
SELO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SELO - selenoprotein O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031454.1 | 5645 | Intron | NP_113642.1 |
TUBGCP6 - tubulin gamma complex associated protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020461.3 | 5645 | Silent Mutation | CGG,CGT | R,R 1711 | NP_065194.2 |