Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATTTTCTCCTGCGTTGGCTCCG[A/C]GGTGAGGGAAGAGGGGCTGCGGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607558 | ||||||||||||||||||||
Literature Links: |
SEC14L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEC14L2 - SEC14 like lipid binding 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204204.2 | 305 | Silent Mutation | CGA,CGC | R,R 43 | NP_001191133.1 | |
NM_001291932.1 | 305 | Intron | NP_001278861.1 | |||
NM_012429.4 | 305 | Silent Mutation | CGA,CGC | R,R 43 | NP_036561.1 | |
NM_033382.2 | 305 | Silent Mutation | CGA,CGC | R,R 43 | NP_203740.1 |