Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACACACGTTATCATTTGGGAGAC[C/T]GGGTGGAAGGTATGCAAGATCTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615117 MIM: 610690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf88 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf88 - chromosome 2 open reading frame 88 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIBCH - 3-hydroxyisobutyryl-CoA hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014362.3 | Intron | NP_055177.2 | ||||
NM_198047.2 | Intron | NP_932164.1 | ||||
XM_011510953.1 | Intron | XP_011509255.1 | ||||
XM_011510954.1 | Intron | XP_011509256.1 |