Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGGTGCTGTTGCTGAAGGAGGC[C/T]ACGGGGTGCAGTGGGAACTCACGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182098 | ||||||||||||||||||||
Literature Links: |
SCTR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCTR - secretin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002980.2 | 847 | Silent Mutation | GTA,GTG | V,V 416 | NP_002971.2 | |
XM_005263730.4 | 847 | Silent Mutation | GTA,GTG | V,V 238 | XP_005263787.1 | |
XM_011511621.2 | 847 | Silent Mutation | GTA,GTG | V,V 421 | XP_011509923.1 | |
XM_017004670.1 | 847 | Silent Mutation | GTA,GTG | V,V 411 | XP_016860159.1 | |
XM_017004671.1 | 847 | Silent Mutation | GTA,GTG | V,V 385 | XP_016860160.1 | |
XM_017004672.1 | 847 | Silent Mutation | GTA,GTG | V,V 353 | XP_016860161.1 | |
XM_017004673.1 | 847 | Silent Mutation | GTA,GTG | V,V 243 | XP_016860162.1 |
TMEM37 - transmembrane protein 37 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |