Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAGAACCACATCAGCTGGGTAC[A/C]GATTGCCCCCCACCAGGCCACCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C2orf50 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf50 - chromosome 2 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182500.2 | 334 | Silent Mutation | AGA,CGA | R,R 18 | NP_872306.1 | |
XM_006711857.2 | 334 | Silent Mutation | AGA,CGA | R,R 18 | XP_006711920.1 | |
XM_006711858.2 | 334 | Silent Mutation | AGA,CGA | R,R 18 | XP_006711921.1 | |
XM_006711859.3 | 334 | Silent Mutation | AGA,CGA | R,R 18 | XP_006711922.1 |
FLJ33534 - uncharacterized LOC285150 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |