Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGCTGGTCTCCGGGGTTCTGGC[A/C]CCTGCGGTGCTCACAGGTAAGAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612003 MIM: 605991 | ||||||||||||||||||||
Literature Links: |
C2orf82 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf82 - chromosome 2 open reading frame 82 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206895.1 | 178 | Silent Mutation | GCA,GCC | A,A 19 | NP_996778.1 | |
XM_017004080.1 | 178 | Silent Mutation | GCA,GCC | A,A 118 | XP_016859569.1 | |
XM_017004081.1 | 178 | Silent Mutation | GCA,GCC | A,A 19 | XP_016859570.1 | |
XM_017004082.1 | 178 | Silent Mutation | GCA,GCC | A,A 19 | XP_016859571.1 | |
XM_017004083.1 | 178 | Silent Mutation | GCA,GCC | A,A 19 | XP_016859572.1 | |
XM_017004084.1 | 178 | Silent Mutation | GCA,GCC | A,A 19 | XP_016859573.1 |
GIGYF2 - GRB10 interacting GYF protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NGEF - neuronal guanine nucleotide exchange factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |