Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACTCCATAAAAGAAGAAATAAG[C/T]GAGAAATAATTCCCTGCTTTTACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602310 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100505984 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100505984 - uncharacterized LOC100505984 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBMS1 - RNA binding motif single stranded interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002897.4 | Intron | NP_002888.1 | ||||
NM_016836.3 | Intron | NP_058520.1 | ||||
XM_005246737.1 | Intron | XP_005246794.1 | ||||
XM_005246738.2 | Intron | XP_005246795.1 | ||||
XM_005246739.2 | Intron | XP_005246796.1 | ||||
XM_005246740.1 | Intron | XP_005246797.1 | ||||
XM_005246741.3 | Intron | XP_005246798.1 | ||||
XM_017004626.1 | Intron | XP_016860115.1 |