Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCAGAAAACATCCAGGCAGCAG[C/T]TGACGTTGGAGAAACACATGGACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604107 | ||||||||||||||||||||
Literature Links: |
GPR55 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR55 - G protein-coupled receptor 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005683.3 | 1032 | Missense Mutation | AAC,AGC | N,S 280 | NP_005674.2 | |
XM_005246952.3 | 1032 | Missense Mutation | AAC,AGC | N,S 280 | XP_005247009.1 | |
XM_011512175.2 | 1032 | Missense Mutation | AAC,AGC | N,S 280 | XP_011510477.1 | |
XM_011512176.2 | 1032 | Missense Mutation | AAC,AGC | N,S 280 | XP_011510478.1 |
LOC151484 - uncharacterized LOC151484 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |