Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAAGCCTGGCCTACATTGCCAC[C/G]CAAGGACCCTTGCCTCACACCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606587 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPN18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPN18 - protein tyrosine phosphatase, non-receptor type 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142370.1 | 408 | Intron | NP_001135842.1 | |||
NM_014369.3 | 408 | Silent Mutation | ACC,ACG | T,T 104 | NP_055184.2 | |
XM_006712416.3 | 408 | Intron | XP_006712479.1 | |||
XM_006712417.1 | 408 | Silent Mutation | ACC,ACG | T,T 104 | XP_006712480.1 | |
XM_017003798.1 | 408 | Intron | XP_016859287.1 |