Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTACATGAAGTTCCCTTTCCTG[C/T]CAATATTGGTGAGTAAACCAGTAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611479 | ||||||||||||||||||||
Literature Links: |
CCDC121 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC121 - coiled-coil domain containing 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142683.2 | 260 | Intron | NP_001136155.1 | |||
NM_024584.4 | 260 | Intron | NP_078860.2 | |||
XM_005264560.3 | 260 | Intron | XP_005264617.1 |
GPN1 - GPN-loop GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145047.1 | 260 | Missense Mutation | GCC,GTC | A,V 54 | NP_001138519.1 | |
NM_001145048.1 | 260 | UTR 5 | NP_001138520.1 | |||
NM_001145049.1 | 260 | Missense Mutation | CCA,TCA | P,S 7 | NP_001138521.1 | |
NM_007266.3 | 260 | Missense Mutation | GCC,GTC | A,V 80 | NP_009197.2 |
ZNF512 - zinc finger protein 512 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |