Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGGGTTTGATTTTGTTTTGCAG[G/T]ATCTTGTTCAAGATGGGGTGGCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612711 | ||||||||||||||||||||
Literature Links: |
GCC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GCC2 - GRIP and coiled-coil domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181453.3 | 161 | Missense Mutation | GAT,TAT | D,Y 3 | NP_852118.1 | |
XM_006712870.3 | 161 | Missense Mutation | GAT,TAT | D,Y 6 | XP_006712933.1 | |
XM_006712871.1 | 161 | UTR 5 | XP_006712934.1 | |||
XM_006712872.3 | 161 | Missense Mutation | GAT,TAT | D,Y 6 | XP_006712935.1 | |
XM_011512213.2 | 161 | Intron | XP_011510515.1 |