Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCACTCCGCAGTGGTAGCAGGCC[C/T]GAAGGGGCAGGTAACAGCACATACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609292 | ||||||||||||||||||||
Literature Links: |
SPRED2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPRED2 - sprouty related EVH1 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128210.1 | 1143 | Missense Mutation | CAG,CGG | Q,R 395 | NP_001121682.1 | |
NM_181784.2 | 1143 | Missense Mutation | CAG,CGG | Q,R 398 | NP_861449.2 | |
XM_005264200.4 | 1143 | Intron | XP_005264257.2 | |||
XM_005264201.1 | 1143 | Missense Mutation | CAG,CGG | Q,R 201 | XP_005264258.1 | |
XM_005264202.4 | 1143 | Intron | XP_005264259.1 | |||
XM_017003576.1 | 1143 | Missense Mutation | CAG,CGG | Q,R 275 | XP_016859065.1 |