Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCGCCTTCCTCCTCGCCATCGC[C/T]TACTTGATCGTTAAAGGTATTGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616495 MIM: 612941 | ||||||||||||||||||||
Literature Links: |
ARL6IP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL6IP6 - ADP ribosylation factor like GTPase 6 interacting protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152522.5 | 80 | Silent Mutation | GCC,GCT | A,A 128 | NP_689735.1 | |
XM_006712326.3 | 80 | UTR 5 | XP_006712389.1 |
PRPF40A - pre-mRNA processing factor 40 homolog A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |