Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGCAGCGCATTGCGGCTGAACT[C/T]AAATGCCACCCAACGGATGAGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KYNU PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KYNU - kynureninase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032998.1 | 152 | Silent Mutation | CTC,CTT | L,L 20 | NP_001028170.1 | |
NM_001199241.1 | 152 | Silent Mutation | CTC,CTT | L,L 20 | NP_001186170.1 | |
NM_003937.2 | 152 | Silent Mutation | CTC,CTT | L,L 20 | NP_003928.1 | |
XM_011512102.2 | 152 | Intron | XP_011510404.1 | |||
XM_017005217.1 | 152 | Silent Mutation | CTC,CTT | L,L 20 | XP_016860706.1 |