Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGCTGAACGGCCTTAAGAAGC[C/T]CAAGCGGAAAGGCAAGGAGCCGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATOH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATOH8 - atonal bHLH transcription factor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032827.6 | 481 | Missense Mutation | CCC,CTC | P,L 26 | NP_116216.2 | |
XM_006712122.3 | 481 | Missense Mutation | CCC,CTC | P,L 26 | XP_006712185.1 | |
XM_011533139.1 | 481 | Missense Mutation | CCC,CTC | P,L 26 | XP_011531441.1 |