Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCCTCGTCCAGCCGCACACGGC[A/G]ACTCTGCATGAGGTATGCCAGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608860 MIM: 603068 | ||||||||||||||||||||
Literature Links: |
ASTL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASTL - astacin-like metallo-endopeptidase (M12 family) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002036.3 | 912 | Intron | NP_001002036.3 | |||
XM_011511205.2 | 912 | Intron | XP_011509507.1 | |||
XM_011511206.2 | 912 | Intron | XP_011509508.1 | |||
XM_011511207.2 | 912 | Intron | XP_011509509.1 | |||
XM_011511208.2 | 912 | Intron | XP_011509510.1 |
DUSP2 - dual specificity phosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004418.3 | 912 | Missense Mutation | CGC,TGC | R,C 276 | NP_004409.1 | |
XM_017003546.1 | 912 | Missense Mutation | CGC,TGC | R,C 304 | XP_016859035.1 |