Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATACACTGTGGAATCGGAGGTC[A/C]GTTTAGTTGGTGTGGCAAATATGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605771 | ||||||||||||||||||||
Literature Links: |
COX7A2L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COX7A2L - cytochrome c oxidase subunit 7A2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319036.1 | 216 | Missense Mutation | CGG,CTG | R,L 45 | NP_001305965.1 | |
NM_001319037.1 | 216 | Intron | NP_001305966.1 | |||
NM_001319038.1 | 216 | Missense Mutation | CGG,CTG | R,L 45 | NP_001305967.1 | |
NM_001319040.1 | 216 | Intron | NP_001305969.1 | |||
NM_004718.3 | 216 | Intron | NP_004709.2 |