Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGTCCAGAATGGAACAGAACC[G/T]TCATCCCTGCCATTCTTGGACCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114010 MIM: 604024 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATRAID PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATRAID - all-trans retinoic acid induced differentiation factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CAD - carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306079.1 | 597 | Silent Mutation | CCG,CCT | P,P 145 | NP_001293008.1 | |
NM_004341.4 | 597 | Silent Mutation | CCG,CCT | P,P 145 | NP_004332.2 | |
XM_005264555.3 | 597 | Silent Mutation | CCG,CCT | P,P 145 | XP_005264612.1 | |
XM_006712101.2 | 597 | Silent Mutation | CCG,CCT | P,P 145 | XP_006712164.1 |
SLC5A6 - solute carrier family 5 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |