Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAAGCTCATGTTTGGTTCCATC[A/G]GTGATTTTAAGAATTTCATTTAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609552 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FER1L5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FER1L5 - fer-1 like family member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LMAN2L - lectin, mannose binding 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142292.1 | 2295 | UTR 3 | NP_001135764.1 | |||
NM_001322346.1 | 2295 | UTR 3 | NP_001309275.1 | |||
NM_001322347.1 | 2295 | UTR 3 | NP_001309276.1 | |||
NM_001322350.1 | 2295 | UTR 3 | NP_001309279.1 | |||
NM_001322351.1 | 2295 | UTR 3 | NP_001309280.1 | |||
NM_001322352.1 | 2295 | UTR 3 | NP_001309281.1 | |||
NM_001322354.1 | 2295 | UTR 3 | NP_001309283.1 | |||
NM_001322355.1 | 2295 | UTR 3 | NP_001309284.1 | |||
NM_001322356.1 | 2295 | UTR 3 | NP_001309285.1 | |||
NM_030805.3 | 2295 | UTR 3 | NP_110432.1 |