Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGGGCAGTGGGGTGCCCTGCA[C/T]GTGCTGGGGACAGCAGCCCTGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608527 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107986004 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107986004 - uncharacterized LOC107986004 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEU4 - neuraminidase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167599.2 | 930 | Silent Mutation | CAC,CAT | H,H 85 | NP_001161071.1 | |
NM_001167600.2 | 930 | Silent Mutation | CAC,CAT | H,H 72 | NP_001161072.1 | |
NM_001167601.2 | 930 | Silent Mutation | CAC,CAT | H,H 72 | NP_001161073.1 | |
NM_001167602.2 | 930 | Silent Mutation | CAC,CAT | H,H 72 | NP_001161074.1 | |
NM_080741.3 | 930 | Silent Mutation | CAC,CAT | H,H 84 | NP_542779.2 |