Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGTGGCACAACTCTTCCTTCCC[C/T]GTGCACAGCAGGAAAGCTGCCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHRS9 - dehydrogenase/reductase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142270.1 | 369 | Intron | NP_001135742.1 | |||
NM_001142271.1 | 369 | Intron | NP_001135743.1 | |||
NM_001289763.1 | 369 | Intron | NP_001276692.1 | |||
NM_199204.1 | 369 | UTR 5 | NP_954674.1 | |||
XM_011510475.1 | 369 | UTR 5 | XP_011508777.1 |