Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTTCATTAAAGATGCAGTATCC[C/T]TGGTTCACACAAATGCATCTGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605295 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FIGN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FIGN - fidgetin, microtubule severing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321825.1 | 6899 | UTR 3 | NP_001308754.1 | |||
NM_018086.3 | 6899 | UTR 3 | NP_060556.2 | |||
XM_011511389.2 | 6899 | UTR 3 | XP_011509691.1 | |||
XM_017004419.1 | 6899 | UTR 3 | XP_016859908.1 |