Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGGGGCTCACGTTGTGGTCGGC[A/G]GTGCAGTAGGCGATGCGGGCCACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612930 | ||||||||||||||||||||
Literature Links: |
PID1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PID1 - phosphotyrosine interaction domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100818.1 | 709 | Silent Mutation | ACC,ACT | T,T 137 | NP_001094288.1 | |
NM_017933.4 | 709 | Silent Mutation | ACC,ACT | T,T 168 | NP_060403.3 | |
XM_011511384.2 | 709 | Silent Mutation | ACC,ACT | T,T 170 | XP_011509686.1 | |
XM_017004404.1 | 709 | Silent Mutation | ACC,ACT | T,T 225 | XP_016859893.1 | |
XM_017004405.1 | 709 | Silent Mutation | ACC,ACT | T,T 188 | XP_016859894.1 | |
XM_017004406.1 | 709 | Silent Mutation | ACC,ACT | T,T 123 | XP_016859895.1 | |
XM_017004407.1 | 709 | Silent Mutation | ACC,ACT | T,T 88 | XP_016859896.1 |