Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCTCACTCCATCAGGCCTTTC[C/T]GGGAACTGCTTTGAAATATTCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CREG2 - cellular repressor of E1A stimulated genes 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153836.3 | 908 | Missense Mutation | AGA,GGA | R,G 288 | NP_722578.1 | |
XM_011510777.2 | 908 | Intron | XP_011509079.1 | |||
XM_017003565.1 | 908 | Intron | XP_016859054.1 |