Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACGCCCAAGAGATGCTGGGTCT[C/T]GTCCACTAGCACCAGCCTGTGTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606530 MIM: 604976 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP27A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP27A1 - cytochrome P450 family 27 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR9500 - microRNA 9500 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKAG3 - protein kinase AMP-activated non-catalytic subunit gamma 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017431.2 | 1397 | Missense Mutation | NP_059127.2 | |||
XM_017004343.1 | 1397 | Nonsense Mutation | XP_016859832.1 |