Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCGTAGGCGCCACGCTCATCCC[A/G]AGGCAGAACCTGCTCCAGCCGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607386 MIM: 606010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT172 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFT172 - intraflagellar transport 172 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015662.2 | 5174 | Missense Mutation | CGG,TGG | R,W 1664 | NP_056477.1 | |
XM_005264254.1 | 5174 | Missense Mutation | CGG,TGG | R,W 1642 | XP_005264311.1 | |
XM_006711986.3 | 5174 | Missense Mutation | CGG,TGG | R,W 1643 | XP_006712049.1 | |
XM_006711987.1 | 5174 | Missense Mutation | CGG,TGG | R,W 1631 | XP_006712050.1 | |
XM_011532757.2 | 5174 | Missense Mutation | CGG,TGG | R,W 1437 | XP_011531059.1 | |
XM_011532758.1 | 5174 | Intron | XP_011531060.1 | |||
XM_011532759.2 | 5174 | Missense Mutation | CGG,TGG | R,W 1144 | XP_011531061.1 | |
XM_011532760.2 | 5174 | Missense Mutation | CGG,TGG | R,W 1019 | XP_011531062.1 | |
XM_017003790.1 | 5174 | Missense Mutation | CGG,TGG | R,W 1621 | XP_016859279.1 | |
XM_017003791.1 | 5174 | Missense Mutation | CGG,TGG | R,W 1437 | XP_016859280.1 | |
XM_017003792.1 | 5174 | Intron | XP_016859281.1 | |||
XM_017003793.1 | 5174 | Missense Mutation | CGG,TGG | R,W 1043 | XP_016859282.1 | |
XM_017003794.1 | 5174 | Missense Mutation | CGG,TGG | R,W 1043 | XP_016859283.1 | |
XM_017003795.1 | 5174 | Missense Mutation | CGG,TGG | R,W 975 | XP_016859284.1 |
KRTCAP3 - keratinocyte associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168364.1 | 5174 | Intron | NP_001161836.1 | |||
NM_001321325.1 | 5174 | Intron | NP_001308254.1 | |||
NM_173853.3 | 5174 | Intron | NP_776252.2 |
NRBP1 - nuclear receptor binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |