Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTATTGCTTTATTTACCTTCA[A/G]AGTATTCCAGTAGTGGACTGCCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605683 MIM: 613334 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAZ2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAZ2B - bromodomain adjacent to zinc finger domain 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MARCH7 - membrane associated ring-CH-type finger 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282805.1 | Intron | NP_001269734.1 | ||||
NM_001282806.1 | Intron | NP_001269735.1 | ||||
NM_001282807.1 | Intron | NP_001269736.1 | ||||
NM_022826.3 | Intron | NP_073737.1 | ||||
XM_005246773.2 | Intron | XP_005246830.1 | ||||
XM_005246774.1 | Intron | XP_005246831.1 | ||||
XM_017004718.1 | Intron | XP_016860207.1 | ||||
XM_017004719.1 | Intron | XP_016860208.1 | ||||
XM_017004720.1 | Intron | XP_016860209.1 | ||||
XM_017004721.1 | Intron | XP_016860210.1 |