Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTGTGCCACACCCCGGCTGTTT[A/C]CAGATGACCCACGGGAAGGGCAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605286 | ||||||||||||||||||||
Literature Links: |
CAPN10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPN10 - calpain 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023083.3 | 365 | Missense Mutation | ACA,CCA | T,P 57 | NP_075571.1 | |
NM_023085.3 | 365 | Missense Mutation | ACA,CCA | T,P 57 | NP_075573.2 |
CAPN10-AS1 - CAPN10 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |