Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGACCCTCAACAAAGTCTTCATA[G/T]GTGGCCTGGTCCTTGTTGTTGGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYL1 - myosin light chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_079420.2 | 323 | Silent Mutation | ACA,ACC | T,T 128 | NP_524144.1 | |
NM_079422.2 | 323 | Silent Mutation | ACA,ACC | T,T 84 | NP_524146.1 |