Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGAAGGTCTAGCCATCTGTGGTT[C/T]GGATCGCCGAGGTAGCGCACGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606376 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHST10 - carbohydrate sulfotransferase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004854.4 | 1463 | Silent Mutation | CCA,CCG | P,P 247 | NP_004845.1 | |
XM_011512207.1 | 1463 | Silent Mutation | CCA,CCG | P,P 247 | XP_011510509.1 | |
XM_011512208.1 | 1463 | Silent Mutation | CCA,CCG | P,P 247 | XP_011510510.1 | |
XM_011512210.1 | 1463 | Silent Mutation | CCA,CCG | P,P 247 | XP_011510512.1 | |
XM_011512211.1 | 1463 | Silent Mutation | CCA,CCG | P,P 247 | XP_011510513.1 | |
XM_011512212.1 | 1463 | Silent Mutation | CCA,CCG | P,P 247 | XP_011510514.1 | |
XM_017005380.1 | 1463 | Silent Mutation | CCA,CCG | P,P 247 | XP_016860869.1 | |
XM_017005381.1 | 1463 | Silent Mutation | CCA,CCG | P,P 247 | XP_016860870.1 | |
XM_017005382.1 | 1463 | Silent Mutation | CCA,CCG | P,P 247 | XP_016860871.1 | |
XM_017005383.1 | 1463 | Silent Mutation | CCA,CCG | P,P 247 | XP_016860872.1 |