Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTCTCACCTGTGTTTGTTCTTC[A/T]GGAAGGAGATCAAATATAGCTTCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191760 MIM: 614633 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UGP2 - UDP-glucose pyrophosphorylase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS54 - VPS54, GARP complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005739.1 | 2845 | Silent Mutation | CCA,CCT | P,P 894 | NP_001005739.1 | |
NM_016516.2 | 2845 | Silent Mutation | CCA,CCT | P,P 906 | NP_057600.2 | |
XM_006712029.3 | 2845 | Silent Mutation | CCA,CCT | P,P 870 | XP_006712092.1 | |
XM_017004316.1 | 2845 | Silent Mutation | CCA,CCT | P,P 788 | XP_016859805.1 |