Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGATCCCTTCTTTGCTCATGTC[C/T]GCAGGGGAGGCTGCTCTATCTGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607386 MIM: 606010 | ||||||||||||||||||||
Literature Links: |
IFT172 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFT172 - intraflagellar transport 172 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015662.2 | 569 | Intron | NP_056477.1 | |||
XM_005264254.1 | 569 | Intron | XP_005264311.1 | |||
XM_006711986.3 | 569 | Intron | XP_006712049.1 | |||
XM_006711987.1 | 569 | Intron | XP_006712050.1 | |||
XM_011532757.2 | 569 | Intron | XP_011531059.1 | |||
XM_011532758.1 | 569 | Intron | XP_011531060.1 | |||
XM_011532759.2 | 569 | Intron | XP_011531061.1 | |||
XM_011532760.2 | 569 | Intron | XP_011531062.1 | |||
XM_017003790.1 | 569 | Intron | XP_016859279.1 | |||
XM_017003791.1 | 569 | Intron | XP_016859280.1 | |||
XM_017003792.1 | 569 | Intron | XP_016859281.1 | |||
XM_017003793.1 | 569 | Intron | XP_016859282.1 | |||
XM_017003794.1 | 569 | Intron | XP_016859283.1 | |||
XM_017003795.1 | 569 | Intron | XP_016859284.1 |
KRTCAP3 - keratinocyte associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168364.1 | 569 | Silent Mutation | TCC,TCT | S,S 174 | NP_001161836.1 | |
NM_001321325.1 | 569 | Silent Mutation | TCC,TCT | S,S 174 | NP_001308254.1 | |
NM_173853.3 | 569 | Silent Mutation | TCC,TCT | S,S 174 | NP_776252.2 |
NRBP1 - nuclear receptor binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |