Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCAGGAGCAGGTTGGTCTTGAC[A/G]TGCGGATCATTGAACTTAGGGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604333 MIM: 601664 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIAO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CIAO1 - cytosolic iron-sulfur assembly component 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNRNP200 - small nuclear ribonucleoprotein U5 subunit 200 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014014.4 | 6626 | Silent Mutation | CAC,CAT | H,H 1888 | NP_054733.2 | |
XM_017003603.1 | 6626 | Silent Mutation | CAC,CAT | H,H 1888 | XP_016859092.1 |