Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCAAAAATGGAAGATTTTAACA[G/T]GGAGAGTGACAGCTTTGAAGATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146928 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXCR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXCR2 - C-X-C motif chemokine receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168298.1 | 98 | Missense Mutation | AGG,ATG | R,M 6 | NP_001161770.1 | |
NM_001557.3 | 98 | Missense Mutation | AGG,ATG | R,M 6 | NP_001548.1 | |
XM_005246530.3 | 98 | Intron | XP_005246587.1 | |||
XM_017003990.1 | 98 | Missense Mutation | AGG,ATG | R,M 6 | XP_016859479.1 | |
XM_017003991.1 | 98 | Missense Mutation | AGG,ATG | R,M 6 | XP_016859480.1 | |
XM_017003992.1 | 98 | Missense Mutation | AGG,ATG | R,M 6 | XP_016859481.1 |