Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGCTGTGCTCCAGGTCGGCCT[A/G]GGCCCCTGCGCCGTCATCGGCAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616797 MIM: 176830 | ||||||||||||||||||||
Literature Links: |
EFR3B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFR3B - EFR3 homolog B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POMC - proopiomelanocortin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000939.3 | 1008 | Nonsense Mutation | CAG,TAG | Q,* 202 | NP_000930.1 | |
NM_001035256.2 | 1008 | Nonsense Mutation | CAG,TAG | Q,* 202 | NP_001030333.1 | |
NM_001319204.1 | 1008 | Nonsense Mutation | CAG,TAG | Q,* 202 | NP_001306133.1 | |
NM_001319205.1 | 1008 | Nonsense Mutation | CAG,TAG | Q,* 202 | NP_001306134.1 |