Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTTCTAGCAATGAAGCTTGG[A/C]GATACATGGGTGGCTTTGCAAAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603839 | ||||||||||||||||||||
Literature Links: |
NDUFB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFB3 - NADH:ubiquinone oxidoreductase subunit B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257102.1 | 477 | Silent Mutation | AGA,CGA | R,R 52 | NP_001244031.1 | |
NM_002491.2 | 477 | Silent Mutation | AGA,CGA | R,R 52 | NP_002482.1 | |
XM_011511230.2 | 477 | Intron | XP_011509532.1 | |||
XM_017004186.1 | 477 | Silent Mutation | AGA,CGA | R,R 52 | XP_016859675.1 |