Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTTTTGTCATCTGTTTTGTTCC[A/G]GCAATTCAGTGAGGTATGCTTCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MIR4765 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4765 - microRNA 4765 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC27 - tetratricopeptide repeat domain 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193509.1 | 656 | Missense Mutation | CAG,CGG | Q,R 78 | NP_001180438.1 | |
NM_017735.4 | 656 | Missense Mutation | CAG,CGG | Q,R 128 | NP_060205.3 | |
XM_005264416.1 | 656 | Missense Mutation | CAG,CGG | Q,R 128 | XP_005264473.1 | |
XM_011532958.1 | 656 | Missense Mutation | CAG,CGG | Q,R 128 | XP_011531260.1 |