Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAACATAAGCAGCAAGTCATCT[C/T]CCTTCTTCCTGTTCCCCAGGTAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602637 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPP2 - secreted phosphoprotein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006944.2 | Intron | NP_008875.1 | ||||
XM_005246102.3 | Intron | XP_005246159.1 | ||||
XM_011511698.2 | Intron | XP_011510000.1 | ||||
XM_011511699.2 | Intron | XP_011510001.1 | ||||
XM_011511700.2 | Intron | XP_011510002.1 |