Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTAGGGCAAGATCTCCACAGGG[A/C]CACCCTGGAGGGAACAGAGGGGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608860 MIM: 603068 | ||||||||||||||||||||
Literature Links: |
ASTL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASTL - astacin-like metallo-endopeptidase (M12 family) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DUSP2 - dual specificity phosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004418.3 | 601 | Missense Mutation | GGC,GTC | G,V 172 | NP_004409.1 | |
XM_017003546.1 | 601 | Missense Mutation | GGC,GTC | G,V 200 | XP_016859035.1 |