Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCCCTTCATCTCGAGAGGGAAA[A/G]AACTCCTTTGGGGTAAGTAGCAAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606765 | ||||||||||||||||||||
Literature Links: |
TPO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TPO - thyroid peroxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000547.5 | 168 | Missense Mutation | AAA,GAA | K,E 28 | NP_000538.3 | |
NM_001206744.1 | 168 | Missense Mutation | AAA,GAA | K,E 28 | NP_001193673.1 | |
NM_001206745.1 | 168 | Missense Mutation | AAA,GAA | K,E 28 | NP_001193674.1 | |
NM_175719.3 | 168 | Missense Mutation | AAA,GAA | K,E 28 | NP_783650.1 | |
NM_175721.3 | 168 | Missense Mutation | AAA,GAA | K,E 28 | NP_783652.1 | |
NM_175722.3 | 168 | Missense Mutation | AAA,GAA | K,E 28 | NP_783653.1 | |
XM_011510379.2 | 168 | Missense Mutation | AAA,GAA | K,E 28 | XP_011508681.1 | |
XM_011510380.2 | 168 | Missense Mutation | AAA,GAA | K,E 28 | XP_011508682.1 | |
XM_011510381.2 | 168 | Missense Mutation | AAA,GAA | K,E 28 | XP_011508683.1 |