Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACGAATAATTTTTAGTGTGTGTC[C/T]AATGCAATATTTGGGACATCCTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102545 MIM: 601465 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTG2 - actin, gamma 2, smooth muscle, enteric | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DGUOK - deoxyguanosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318859.1 | Intron | NP_001305788.1 | ||||
NM_001318860.1 | Intron | NP_001305789.1 | ||||
NM_001318861.1 | Intron | NP_001305790.1 | ||||
NM_001318862.1 | Intron | NP_001305791.1 | ||||
NM_001318863.1 | Intron | NP_001305792.1 | ||||
NM_080916.2 | Intron | NP_550438.1 | ||||
NM_080918.2 | Intron | NP_550440.1 | ||||
XM_011532647.2 | Intron | XP_011530949.1 |