Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATCAGATCAGGAACCAGAATGCC[C/T]GGAATAGTGACTTTTCTCGTCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600620 MIM: 616234 | ||||||||||||||||||||
Literature Links: |
FKBP1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FKBP1B - FK506 binding protein 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322963.1 | 1142 | Intron | NP_001309892.1 | |||
NM_001322964.1 | 1142 | Intron | NP_001309893.1 | |||
NM_004116.4 | 1142 | Intron | NP_004107.1 | |||
NM_054033.3 | 1142 | Intron | NP_473374.1 | |||
XM_011532699.2 | 1142 | Intron | XP_011531001.1 | |||
XM_017003593.1 | 1142 | Intron | XP_016859082.1 | |||
XM_017003594.1 | 1142 | Intron | XP_016859083.1 | |||
XM_017003595.1 | 1142 | Intron | XP_016859084.1 | |||
XM_017003596.1 | 1142 | Intron | XP_016859085.1 |
WDCP - WD repeat and coiled coil containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142319.1 | 1142 | Silent Mutation | CCA,CCG | P,P 337 | NP_001135791.1 | |
NM_025203.2 | 1142 | Silent Mutation | CCA,CCG | P,P 337 | NP_079479.1 | |
XM_017005029.1 | 1142 | Silent Mutation | CCA,CCG | P,P 337 | XP_016860518.1 |