Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGGCGGACTCAAGCACCGGCAG[C/T]CCCACAAGCATCGACAAGATCGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01158 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01158 - long intergenic non-protein coding RNA 1158 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC01159 - long intergenic non-protein coding RNA 1159 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POU3F3 - POU class 3 homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006236.2 | 2622 | Silent Mutation | AGC,AGT | S,S 393 | NP_006227.1 |