Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGAACGGGACTTCATCATGCC[A/G]CTGGACTTAGGAGCCAAGGGCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609186 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
D2HGDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
D2HGDH - D-2-hydroxyglutarate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287249.1 | 727 | Silent Mutation | CCA,CCG | P,P 55 | NP_001274178.1 | |
NM_152783.4 | 727 | Silent Mutation | CCA,CCG | P,P 189 | NP_689996.4 | |
XM_011511734.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510036.1 | |
XM_011511735.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510037.1 | |
XM_011511736.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510038.1 | |
XM_011511737.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510039.1 | |
XM_011511738.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510040.1 | |
XM_011511739.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510041.1 | |
XM_011511740.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510042.1 | |
XM_011511741.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510043.1 | |
XM_011511743.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510045.1 | |
XM_011511744.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510046.1 | |
XM_011511745.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510047.1 | |
XM_011511746.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510048.1 | |
XM_011511747.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510049.1 | |
XM_011511749.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510051.1 | |
XM_011511750.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510052.1 | |
XM_011511751.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510053.1 | |
XM_011511752.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510054.1 | |
XM_011511753.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510055.1 | |
XM_011511754.1 | 727 | Silent Mutation | CCA,CCG | P,P 2 | XP_011510056.1 | |
XM_011511756.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510058.1 | |
XM_011511757.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510059.1 | |
XM_011511758.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510060.1 | |
XM_011511759.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510061.1 | |
XM_011511760.2 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_011510062.1 | |
XM_017004827.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_016860316.1 | |
XM_017004828.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_016860317.1 | |
XM_017004829.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_016860318.1 | |
XM_017004830.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_016860319.1 | |
XM_017004831.1 | 727 | Silent Mutation | CCA,CCG | P,P 2 | XP_016860320.1 | |
XM_017004832.1 | 727 | Silent Mutation | CCA,CCG | P,P 189 | XP_016860321.1 |