Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGAGTGTAGTTGTGGGCACCAG[C/T]GGCTGGGGCCACAAAGTCCAACTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604333 MIM: 601664 MIM: 613403 | ||||||||||||||||||||
Literature Links: |
CIAO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CIAO1 - cytosolic iron-sulfur assembly component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004804.2 | 7257 | Intron | NP_004795.1 |
SNRNP200 - small nuclear ribonucleoprotein U5 subunit 200 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014014.4 | 7257 | Missense Mutation | ACT,GCT | T,A 2099 | NP_054733.2 | |
XM_017003603.1 | 7257 | Missense Mutation | ACT,GCT | T,A 2099 | XP_016859092.1 |
TMEM127 - transmembrane protein 127 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |