Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACTAACTCGACAATCTGAGCTGT[C/G]CTGTTTTCCCAGGTGTTAAATATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUL3 - cullin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257197.1 | 4567 | UTR 3 | NP_001244126.1 | |||
NM_001257198.1 | 4567 | UTR 3 | NP_001244127.1 | |||
NM_003590.4 | 4567 | UTR 3 | NP_003581.1 | |||
XM_006712800.2 | 4567 | Intron | XP_006712863.2 | |||
XM_011511994.2 | 4567 | Intron | XP_011510296.1 | |||
XM_011511995.1 | 4567 | Intron | XP_011510297.1 | |||
XM_011511996.2 | 4567 | Intron | XP_011510298.1 |