Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGAGAGGTAGTGACCCCTGAGA[C/T]ACGTGTTGAACCACAAAATTGTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615347 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATAD2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATAD2B - ATPase family, AAA domain containing 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBXN2A - UBX domain protein 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181713.3 | Intron | NP_859064.2 | ||||
XM_005264166.4 | Intron | XP_005264223.1 | ||||
XM_005264167.4 | Intron | XP_005264224.1 | ||||
XM_005264168.4 | Intron | XP_005264225.1 | ||||
XM_005264169.4 | Intron | XP_005264226.1 | ||||
XM_011532633.2 | Intron | XP_011530935.1 | ||||
XM_017003517.1 | Intron | XP_016859006.1 | ||||
XM_017003518.1 | Intron | XP_016859007.1 |