Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATGCTGAATCTTAAGTGTTTGC[C/T]GTTTCTTTCACTGCGAAAGGGGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602188 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPHA4 - EPH receptor A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304536.1 | 4103 | UTR 3 | NP_001291465.1 | |||
NM_001304537.1 | 4103 | UTR 3 | NP_001291466.1 | |||
NM_004438.4 | 4103 | UTR 3 | NP_004429.1 | |||
XM_005246374.2 | 4103 | UTR 3 | XP_005246431.1 |