Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAGCCACTTGGATTGGTTCTTC[C/T]GTAAACTGTTTCTTAAGCGTCCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
COPS9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COPS9 - COP9 signalosome subunit 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163424.1 | 985 | Intron | NP_001156896.1 | |||
NM_138336.1 | 985 | Missense Mutation | CAG,CGG | Q,R 187 | NP_612209.1 | |
XM_017003411.1 | 985 | Missense Mutation | CAG,CGG | Q,R 177 | XP_016858900.1 |